Merck
Tetraspanin 8 is an interactor of the metalloprotease meprin β within tetraspanin-enriched microdomains.
Biological chemistry (2016-05-18)
Frederike Schmidt, Miryam Müller, Johannes Prox, Philipp Arnold, Caroline Schönherr, Claudia Tredup, Petra Minder, Henriette Ebsen, Ottmar Janssen, Wim Annaert, Claus Pietrzik, Dirk Schmidt-Arras, Erwin E Sterchi, Christoph Becker-Pauly
ABSTRAKT
Meprin β is a dimeric type I transmembrane protein and acts as an ectodomain sheddase at the cell surface. It was shown that meprin β cleaves the amyloid precursor protein (APP), thereby releasing neurotoxic amyloid β peptides and implicating a role of meprin β in Alzheimer's disease. In order to identify non-proteolytic regulators of meprin β, we performed a split ubiquitin yeast two-hybrid screen using a small intestinal cDNA library. In this screen we identified tetraspanin 8 (TSPAN8) as interaction partner for meprin β. Since several members of the tetraspanin family were described to interact with metalloproteases thereby affecting their localization and/or activity, we hypothesized similar functions of TSPAN8 in the regulation of meprin β. We employed cell biological methods to confirm direct binding of TSPAN8 to meprin β. Surprisingly, we did not observe an effect of TSPAN8 on the catalytic activity of meprin β nor on the specific cleavage of its substrate APP. However, both proteins were identified being present in tetraspanin-enriched microdomains. Therefore we hypothesize that TSPAN8 might be important for the orchestration of meprin β at the cell surface with impact on certain proteolytic processes that have to be further identified.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution