Merck
Virulence potential of commensal multidrug resistant Escherichia coli isolated from poultry in Brazil.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases (2018-08-03)
Joseane Cristina Ferreira, Rafael Antonio Casarin Penha Filho, Ana Paula Yorika Kuaye, Leonardo Neves Andrade, Yung-Fu Chang, Ana Lúcia Costa Darini
ABSTRAKT
There is an increasing number of reports worldwide about multidrug resistance (MDR) with potential of ExPEC in commensal E. coli. The present study evaluated the potential ExPEC in selected 44 MDR E.coli isolates, collected from livestock. ExPEC isolates were characterized by analysis of five main groups of virulence genes (papA and/or papC, sfa and/or foc, afa and/or dra, kpsMT II and iutA). We also determined the increased virulence potential analyzing other 29 virulence genes, the epidemiology of these isolates. Additionally, fifteen ExPEC isolates were selected to evaluate the adhesion and invasion capacity in vitro using Caco-2 cells. Based on the analysis of the five main virulence genes, 72.7% (32/44) strains were classified as ExPEC. The presence of each gene was iutA 88.6%, KpsMT II 70.4%, papC 25%, sfa/focDE 4.5%; afa/draBC genes were not found. All E. coli isolates were classified into: phylogenetic groups A (34%), B1 (10%), B2 (20%), and D (36%). MLST revealed 7 different STs among isolates, including a new ST identified (ST5687). The in vitro assay in Caco-2 cells showed that all isolates were capable to adhere or invade the epithelial cells, although this occurred at variable levels. The ExPEC isolate LO122 reached similar levels of invasion to the positive control strain Salmonella Typhimurium LT2. These results showed that the apparently commensal microbiota of poultry harbors MDR ExPEC isolates with high adhesion and invasion potential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide solution