Merck
Association of 5-hydroxytryptamine (serotonin) receptor 4 (5-HTR4) gene polymorphisms with asthma.
Respirology (Carlton, Vic.) (2011-03-09)
Tae-Hoon Kim, Sung-Hye An, Ji-Yeon Cha, Eun-Kyong Shin, Ji-Yeon Lee, Sang-Hyuk Yoon, Young-Mok Lee, Soo-Taek Uh, Sung-Woo Park, Jong-Sook Park, Young-Hoon Kim, Jae-Sung Choi, Soo-Ok Lee, Byung-Lae Park, Hyung-Doo Shin, Choon-Sik Park
ABSTRAKT
The neurotransmitter, 5-hydroxytryptamine, acts as an immunomodulator by stimulating the release of inflammatory cytokines and regulating the function of dendritic cells and monocytes. The 5-hydroxytryptamine receptor 4 (HTR4) gene is located in a region previously linked to an increased risk of asthma and atopy. The aim of this study was to investigate the association between HTR4 and asthma. Thirty-two single nucleotide polymorphisms (SNP) in HTR4 were investigated by direct sequencing of 24 DNA samples from unrelated Korean subjects. The 32 genetic variants comprised 22 intronic SNP, two SNP in the 3'-untranslated region (exon 7) and eight SNP in the 3'-downstream region. Logistic regression analysis showed that two intronic polymorphisms were significantly associated with the risk of asthma. Two minor HTR4 alleles, +142828G>A and +122769G>A, occurred at significantly higher frequencies in the asthmatic group than in the healthy control group (49.59% vs 42.29%, P=0.003, and 47.99% vs 40.35%, P=0.008, respectively), and these differences remained significant after correction for multiple testing (P=0.05, dominant mode of inheritance; and P=0.03, dominant mode, respectively). Haplotype analysis revealed three haplotype blocks. The frequency of haplotype 1 in block 2 was significantly higher in asthmatics (P=0.003, dominant mode), whereas the frequency of haplotype 4 in block 3 was significantly lower in asthmatics (P=0.0009, dominant mode). SNP and haplotypes of the HTR4 gene were associated with the asthma phenotype and genetic variation of HTR4 may affect susceptibility to the development of asthma.