- Recyclable hypervalent iodine(III) reagent iodosodilactone as an efficient coupling reagent for direct esterification, amidation, and peptide coupling.
Recyclable hypervalent iodine(III) reagent iodosodilactone as an efficient coupling reagent for direct esterification, amidation, and peptide coupling.
Organic letters (2012-06-08)
Jun Tian, Wen-Chao Gao, Dong-Mei Zhou, Chi Zhang
PMID22671301
ABSTRAKT
A hypervalent iodine(III) reagent plays a novel role as an efficient coupling reagent to promote the direct condensation between carboxylic acids and alcohols or amines to provide esters, macrocyclic lactones, amides, as well as peptides without racemization. The regeneration of iodosodilactone (1) can also be readily achieved. The intermediate acyloxyphosphonium ion C from the activation of a carboxylic acid is thought to be involved in the present esterification reaction.