Merck
CdSe Quantum Dots in Human Models Derived from ALS Patients: Characterization, Nuclear Penetration Studies and Multiplexing.
Nanomaterials (Basel, Switzerland) (2021-04-04)
Carlota Tosat-Bitrián, Alicia Avis-Bodas, Gracia Porras, Daniel Borrego-Hernández, Alberto García-Redondo, Angeles Martín-Requero, Valle Palomo
ABSTRAKT
CdSe quantum dots (QDs) are valuable tools for deciphering molecular mechanisms in cells. Their conjugation with antibodies offers a unique staining source with optimal characteristics, including increased photostability and narrow emission spectra, allowing for improved multiplexing capabilities using a single excitation source. In combination with pathology models derived from patients, they have great potential to contribute to quantitative molecular profiling and promote personalized medicine. However, the commercial availability of diverse CdSe QDs is still limited and characterization techniques must be performed to these materials or the conjugates developed in the lab to assure a proper function and reproducibility. Furthermore, while there is significant data of QDs experiments in cell lines, the literature with primary human cells is scarce, and QD behavior in these systems may be different. Rigorous characterization data of commercially available QDs and their conjugates with biomolecules of interest is needed in order to establish their potential for target labelling and expand their use among research labs. Here we compare the characterization and labelling performance of different QD conjugates in SH-SY5Y cell line, fibroblasts and immortalized lymphocytes derived from amyotrophic lateral sclerosis patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Casein solution from bovine milk, 5% in water
Sigma-Aldrich
Protein A from Staphylococcus aureus, Soluble, essentially salt-free, lyophilized powder, extracellular
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Triton X-100 solution, BioUltra, Molecular Biology, ~10% in H2O