Merck
Identification of a GrgA-Euo-HrcA Transcriptional Regulatory Network in Chlamydia.
mSystems (2021-08-04)
Wurihan Wurihan, Yi Zou, Alec M Weber, Korri Weldon, Yehong Huang, Xiaofeng Bao, Chengsheng Zhu, Xiang Wu, Yaqun Wang, Zhao Lai, Huizhou Fan
ABSTRAKT
Chlamydia trachomatis is an obligate intracellular bacterium whose unique developmental cycle consists of an infectious elementary body and a replicative reticulate body. Progression of this developmental cycle requires temporal control of the transcriptome. In addition to the three chlamydial sigma factors (σ66, σ28, and σ54) that recognize promoter sequences of genes, chlamydial transcription factors are expected to play crucial roles in transcriptional regulation. Here, we investigate the function of GrgA, a Chlamydia-specific transcription factor, in C. trachomatis transcriptomic expression. We show that 10 to 30 min of GrgA overexpression induces 13 genes, which likely comprise the direct regulon of GrgA. Significantly, σ66-dependent genes that code for two important transcription repressors are components of the direct regulon. One of these repressors is Euo, which prevents the expression of late genes during early phases. The other is HrcA, which regulates molecular chaperone expression and controls stress response. The direct regulon also includes a σ28-dependent gene that codes for the putative virulence factor PmpI. Furthermore, overexpression of GrgA leads to decreased expression of almost all tRNAs. Transcriptomic studies suggest that GrgA, Euo, and HrcA have distinct but overlapping indirect regulons. These findings, together with temporal expression patterns of grgA, euo, and hrcA, indicate that a transcriptional regulatory network of these three transcription factors plays critical roles in C. trachomatis growth and development. IMPORTANCE Chlamydia trachomatis is the most prevalent sexually transmitted bacterial pathogen worldwide and is a leading cause of preventable blindness in underdeveloped areas as well as some developed countries. Chlamydia carries genes that encode a limited number of known transcription factors. While Euo is thought to be critical for early chlamydial development, the functions of GrgA and HrcA in the developmental cycle are unclear. Activation of euo and hrcA immediately following GrgA overexpression indicates that GrgA functions as a master transcriptional regulator. In addition, by broadly inhibiting tRNA expression, GrgA serves as a key regulator of chlamydial protein synthesis. Furthermore, by upregulating pmpI, GrgA may act as an upstream virulence determinant. Finally, genes coregulated by GrgA, Euo, and HrcA likely play critical roles in chlamydial growth and developmental control.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
TRI Reagent®