Merck
The amino terminal sequences of acid proteases-human pepsin and gastricsin and the protease of Rhizopus chinensis.
Biochemical and biophysical research communications (1975-04-21)
P Sepulveda, K W Jackson, J Tang
PMID236753
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protease from Rhizopus sp.