- Extraction of Soluble and Insoluble Protein Fractions from Mouse Brains and Spinal Cords.
Extraction of Soluble and Insoluble Protein Fractions from Mouse Brains and Spinal Cords.
Bio-protocol (2017-08-05)
Oliver Wirths
PMID34541150
ABSTRAKT
The current protocol details the preparation of soluble and insoluble protein lysates from mouse brain or spinal cord samples. In detail, tissue homogenization and sequential protein extraction are described. This procedure yields soluble and insoluble protein extracts that can be further processed in down-stream applications like ELISA or Western blotting.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Roche
cOmplete™™ Mini, Koktajl inhibitorów proteaz bez EDTA, Tablets provided in EASYpacks