Merck
Valorization of Greenhouse Horticulture Waste from a Biorefinery Perspective.
Foods (Basel, Switzerland) (2021-05-01)
Antonio D Moreno, Aleta Duque, Alberto González, Ignacio Ballesteros, María José Negro
ABSTRAKT
Greenhouse cultivation and harvesting generate considerable amounts of organic waste, including vegetal waste from plants and discarded products. This study evaluated the residues derived from tomato cultivation practices in Almería (Spain) as sugar-rich raw materials for biorefineries. First, lignocellulose-based residues were subjected to an alkali-catalyzed extrusion process in a twin-screw extruder (100 °C and 6-12% (w/w) NaOH) to assess maximum sugar recovery during the subsequent enzymatic hydrolysis step. A high saccharification yield was reached when using an alkali concentration of 12% (w/w), releasing up to 81% of the initial glucan. Second, the discarded tomato residue was crushed and centrifuged to collect both the juice and the pulp fractions. The juice contained 39.4 g of sugars per 100 g of dry culled tomato, while the pulp yielded an extra 9.1 g of sugars per 100 g of dry culled tomato after an enzymatic hydrolysis process. The results presented herein show the potential of using horticulture waste as an attractive sugar source for biorefineries, including lignocellulose-based residues when effective fractionation processes, such as reactive extrusion technology, are available.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cellulase, enzyme blend