Merck
Atezolizumab with bevacizumab, paclitaxel and carboplatin was effective for patients with SMARCA4-deficient thoracic sarcoma.
Immunotherapy (2021-05-26)
Hayato Kawachi, Kei Kunimasa, Yoji Kukita, Harumi Nakamura, Keiichiro Honma, Takahisa Kawamura, Takako Inoue, Motohiro Tamiya, Hanako Kuhara, Kazumi Nishino, Yu Mizote, Takashi Akazawa, Hideaki Tahara, Toru Kumagai
ABSTRAKT
SMARCA4-deficient thoracic sarcoma (DTS) is a recently noted progressive thoracic malignancy. We recently experienced three cases of SMARCA4-DTS who were treated with atezolizumab in combination with bevacizumab, paclitaxel and carboplatin (ABCP) as the first-line therapy. Immunohistopathological analysis revealed absent expression of SMARCA4 in all cases. The tumor mutational burden was over 11/Mb and mutations in SMARCA4 and TP53 were detected in all three cases. Partial response to ABCP treatment was observed in all three cases, with a progression-free survival of approximately 6 months or longer and a continuous response of 1 year or longer in one case. The first-line ABCP treatment demonstrated durable efficacy in SMARCA4-DTS regardless of the degree of PD-L1 expression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-SMARCA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit