Merck
  • Induced pluripotent stem cell line (INSAi001-A) from a Gaucher disease type 3 patient compound heterozygote for mutations in the GBA1 gene.
Induced pluripotent stem cell line (INSAi001-A) from a Gaucher disease type 3 patient compound heterozygote for mutations in the GBA1 gene.
Stem cell research (2019-11-05)
Ana Joana Duarte, Diogo Ribeiro, Renato Santos, Luciana Moreira, José Bragança, Olga Amaral
ABSTRAKT
Gaucher Disease (GD) type 3 is a neurological form of a multisystemic autosomal recessive disorder belonging to the group of lysosomal storage diseases. Causal mutations in the glucocerebrosidase 1 (GBA1) commonly lead to abnormal protein and GD, heterozygosity is a genetic risk factor for Parkinson's disease. This work describes the use of a non-integrative approach using Sendai Virus delivery to establish induced Pluripotent Stem Cells (iPSCs) from fibroblasts from a GD type 3 patient. Differentiation of iPSCs can be employed to generate a variety of complex cell types with a high degree of genetic complexity that would otherwise be unattainable.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, clone 7F7.1, from mouse
Sigma-Aldrich
Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, clone 7F9.2, from mouse