Merck
Trauma-induced heme release increases susceptibility to bacterial infection.
JCI insight (2021-09-15)
Ghee Rye Lee, David Gallo, Rodrigo W Alves de Souza, Shilpa Tiwari-Heckler, Eva Csizmadia, James D Harbison, Sidharth Shankar, Valerie Banner-Goodspeed, Michael B Yaffe, Maria Serena Longhi, Carl J Hauser, Leo E Otterbein
ABSTRAKT
Infection is a common complication of major trauma that causes significantly increased morbidity and mortality. The mechanisms, however, linking tissue injury to increased susceptibility to infection remain poorly understood. To study this relationship, we present a potentially novel murine model in which a major liver crush injury is followed by bacterial inoculation into the lung. We find that such tissue trauma both impaired bacterial clearance and was associated with significant elevations in plasma heme levels. While neutrophil (PMN) recruitment to the lung in response to Staphylococcus aureus was unchanged after trauma, PMN cleared bacteria poorly. Moreover, PMN show > 50% less expression of TLR2, which is responsible, in part, for bacterial recognition. Administration of heme effectively substituted for trauma. Finally, day 1 trauma patients (n = 9) showed similar elevations in free heme compared with that seen after murine liver injury, and circulating PMN showed similar TLR2 reduction compared with volunteers (n = 6). These findings correlate to high infection rates.