Merck
HDAC6 inhibition restores TDP-43 pathology and axonal transport defects in human motor neurons with TARDBP mutations.
The EMBO journal (2021-03-12)
Raheem Fazal, Steven Boeynaems, Ann Swijsen, Mathias De Decker, Laura Fumagalli, Matthieu Moisse, Joni Vanneste, Wenting Guo, Ruben Boon, Thomas Vercruysse, Kristel Eggermont, Bart Swinnen, Jimmy Beckers, Donya Pakravan, Tijs Vandoorne, Pieter Vanden Berghe, Catherine Verfaillie, Ludo Van Den Bosch, Philip Van Damme
ABSTRAKT
TDP-43 is the major component of pathological inclusions in most ALS patients and in up to 50% of patients with frontotemporal dementia (FTD). Heterozygous missense mutations in TARDBP, the gene encoding TDP-43, are one of the common causes of familial ALS. In this study, we investigate TDP-43 protein behavior in induced pluripotent stem cell (iPSC)-derived motor neurons from three ALS patients with different TARDBP mutations, three healthy controls and an isogenic control. TARDPB mutations induce several TDP-43 changes in spinal motor neurons, including cytoplasmic mislocalization and accumulation of insoluble TDP-43, C-terminal fragments, and phospho-TDP-43. By generating iPSC lines with allele-specific tagging of TDP-43, we find that mutant TDP-43 initiates the observed disease phenotypes and has an altered interactome as indicated by mass spectrometry. Our findings also indicate that TDP-43 proteinopathy results in a defect in mitochondrial transport. Lastly, we show that pharmacological inhibition of histone deacetylase 6 (HDAC6) restores the observed TDP-43 pathologies and the axonal mitochondrial motility, suggesting that HDAC6 inhibition may be an interesting therapeutic target for neurodegenerative disorders linked to TDP-43 pathology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology