Merck
Cyclic stretching boosts microRNA-499 to regulate Bcl-2 via microRNA-208a in atrial fibroblasts.
Journal of cellular and molecular medicine (2021-02-20)
Su-Kiat Chua, Bao-Wei Wang, Ying-Ju Yu, Wei-Jen Fang, Chiu-Mei Lin, Kou-Gi Shyu
ABSTRAKT
MicroRNAs that modulate transcription can regulate other microRNAs and are also up-regulated under pathological stress. MicroRNA-499 (miR-499), microRNA-208a (miR-208a) and B-cell lymphoma 2 (Bcl-2) play roles in cardiovascular diseases, such as direct reprogramming of cardiac fibroblast into cardiomyocyte and cardiomyocyte apoptosis. Whether miR208a, miR499 and Bcl-2 were critical regulators in cardiac fibroblast apoptosis under mechanical stretching conditions in human cardiac fibroblasts-adult atrial (HCF-aa) was investigated. Using negative pressure, HCF-aa grown on a flexible membrane base were cyclically stretched to 20% of their maximum elongation. In adult rats, an aortocaval shunt was used to create an in vivo model of volume overload. MiR208a was up-regulated early by stretching and returned to normal levels with longer stretching cycles, whereas the expression of miR499 and Bcl-2 was up-regulated by longer stretching times. Pre-treatment with antagomir-499 reversed the miR-208a down-regulation, whereas Bcl-2 expression could be suppressed by miR-208a overexpression. In the HCF-aa under stretching for 1 h, miR-499 overexpression decreased pri-miR-208a luciferase activity; this inhibition of pri-miR-208a luciferase activity with stretching was reversed when the miR-499-5p binding site in pri-miR-208a was mutated. The addition of antagomir-208a reversed the Bcl-2-3'UTR suppression from stretching for 1 h. Flow cytometric analysis revealed that pre-treatment with miR-499 or antagomir-208a inhibited cellular apoptosis in stretched HCF-aa. In hearts with volume overload, miR-499 overexpression inhibited myocardial miR-208a expression, whereas Bcl-2 expression could be suppressed by the addition of miR-208a. In conclusion, miR-208a mediated the regulation of miR-499 on Bcl-2 expression in stretched HCF-aa and hearts with volume overload.