Merck
4E-BP2-dependent translation in cerebellar Purkinje cells controls spatial memory but not autism-like behaviors.
Cell reports (2021-04-29)
Mehdi Hooshmandi, Vinh Tai Truong, Eviatar Fields, Riya Elizabeth Thomas, Calvin Wong, Vijendra Sharma, Ilse Gantois, Patricia Soriano Roque, Kleanthi Chalkiadaki, Neil Wu, Anindyo Chakraborty, Soroush Tahmasebi, Masha Prager-Khoutorsky, Nahum Sonenberg, Aparna Suvrathan, Alanna J Watt, Christos G Gkogkas, Arkady Khoutorsky
ABSTRAKT
Recent studies have demonstrated that selective activation of mammalian target of rapamycin complex 1 (mTORC1) in the cerebellum by deletion of the mTORC1 upstream repressors TSC1 or phosphatase and tensin homolog (PTEN) in Purkinje cells (PCs) causes autism-like features and cognitive deficits. However, the molecular mechanisms by which overactivated mTORC1 in the cerebellum engenders these behaviors remain unknown. The eukaryotic translation initiation factor 4E-binding protein 2 (4E-BP2) is a central translational repressor downstream of mTORC1. Here, we show that mice with selective ablation of 4E-BP2 in PCs display a reduced number of PCs, increased regularity of PC action potential firing, and deficits in motor learning. Surprisingly, although spatial memory is impaired in these mice, they exhibit normal social interaction and show no deficits in repetitive behavior. Our data suggest that, downstream of mTORC1/4E-BP2, there are distinct cerebellar mechanisms independently controlling social behavior and memory formation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
RIPA Buffer