- ATP-binding cassette protein ABCF1 couples transcription and genome surveillance in embryonic stem cells through low-complexity domain.
ATP-binding cassette protein ABCF1 couples transcription and genome surveillance in embryonic stem cells through low-complexity domain.
Science advances (2021-10-30)
Eun-Bee Choi, Munender Vodnala, Madeleine Zerbato, Jianing Wang, Jaclyn J Ho, Carla Inouye, Lai Ding, Yick W Fong
PMID34714667
ABSTRAKT
[Figure: see text].
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Przeciwciało anty-trimetylo-histonowe H3 (Lys4), klon 15-10C-E4, rozmiar próbny, clone 15-10C-E4, Upstate®, from rabbit