Merck
Mfge8 attenuates human gastric antrum smooth muscle contractions.
Journal of muscle research and cell motility (2021-06-05)
Wen Li, Ashley Olseen, Yeming Xie, Cristina Alexandru, Andrew Outland, Angela F Herrera, Andrew J Syder, Jill Wykosky, Brian A Perrino
ABSTRAKT
Coordinated gastric smooth muscle contraction is critical for proper digestion and is adversely affected by a number of gastric motility disorders. In this study we report that the secreted protein Mfge8 (milk fat globule-EGF factor 8) inhibits the contractile responses of human gastric antrum muscles to cholinergic stimuli by reducing the inhibitory phosphorylation of the MYPT1 (myosin phosphatase-targeting subunit (1) subunit of MLCP (myosin light chain phosphatase), resulting in reduced LC20 (smooth muscle myosin regulatory light chain (2) phosphorylation. Mfge8 reduced the agonist-induced increase in the F-actin/G-actin ratios of β-actin and γ-actin1. We show that endogenous Mfge8 is bound to its receptor, α8β1 integrin, in human gastric antrum muscles, suggesting that human gastric antrum muscle mechanical responses are regulated by Mfge8. The regulation of gastric antrum smooth muscles by Mfge8 and α8 integrin functions as a brake on gastric antrum mechanical activities. Further studies of the role of Mfge8 and α8 integrin in regulating gastric antrum function will likely reveal additional novel aspects of gastric smooth muscle motility mechanisms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red