Merck
Sialic acid-containing glycolipids mediate binding and viral entry of SARS-CoV-2.
Nature chemical biology (2021-11-11)
Linh Nguyen, Kelli A McCord, Duong T Bui, Kim M Bouwman, Elena N Kitova, Mohamed Elaish, Dhanraj Kumawat, Gour C Daskhan, Ilhan Tomris, Ling Han, Pradeep Chopra, Tzu-Jing Yang, Steven D Willows, Andrew L Mason, Lara K Mahal, Todd L Lowary, Lori J West, Shang-Te Danny Hsu, Tom Hobman, Stephen M Tompkins, Geert-Jan Boons, Robert P de Vries, Matthew S Macauley, John S Klassen
ABSTRAKT
Emerging evidence suggests that host glycans influence severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection. Here, we reveal that the receptor-binding domain (RBD) of the spike (S) protein on SARS-CoV-2 recognizes oligosaccharides containing sialic acid (Sia), with preference for monosialylated gangliosides. Gangliosides embedded within an artificial membrane also bind to the RBD. The monomeric affinities (Kd = 100-200 μM) of gangliosides for the RBD are similar to another negatively charged glycan ligand of the RBD proposed as a viral co-receptor, heparan sulfate (HS) dp2-dp6 oligosaccharides. RBD binding and infection of SARS-CoV-2 pseudotyped lentivirus to angiotensin-converting enzyme 2 (ACE2)-expressing cells is decreased following depletion of cell surface Sia levels using three approaches: sialyltransferase (ST) inhibition, genetic knockout of Sia biosynthesis, or neuraminidase treatment. These effects on RBD binding and both pseudotyped and authentic SARS-CoV-2 viral entry are recapitulated with pharmacological or genetic disruption of glycolipid biosynthesis. Together, these results suggest that sialylated glycans, specifically glycolipids, facilitate viral entry of SARS-CoV-2.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
Neuraminidaza (sialidaza), from Arthrobacter ureafaciens
Roche
Neuraminidaza (sialidaza), from Vibrio cholerae