Merck
Whole mount in situ localization of miRNAs and target mRNA transcripts in plants.
3 Biotech (2019-05-09)
Vibhav Gautam, Archita Singh, Swati Verma, Sharmila Singh, Sourav Chatterjee, Ananda K Sarkar
ABSTRAKT
The functional characterization of miRNAs often involves understanding of their spatiotemporal expression, which mostly relies on reporter-based or in situ hybridization studies. The available in situ localization methods follow separate protocols for pre-hybridization, hybridization, post-hybridization, and detection steps for both miRNA and mRNA transcripts in plants. In this study, we present a single method which can be used for whole mount in situ localization of both miRNAs and mRNAs in different plant tissues. Our modified method provides enhanced sensitivity for the localization of miRNA and their target transcripts. Consequently, a less laborious, time-saving, economic and efficient method has been proposed by the modification of pre-hybridization, hybridization, post-hybridization and detection steps.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
HistoChoice® Clearing Agent, alternative to toluene and xylene