Merck
Optimized Liquid and Gas Phase Fractionation Increases HLA-Peptidome Coverage for Primary Cell and Tissue Samples.
Molecular & cellular proteomics : MCP (2021-08-16)
Susan Klaeger, Annie Apffel, Karl R Clauser, Siranush Sarkizova, Giacomo Oliveira, Suzanna Rachimi, Phuong M Le, Anna Tarren, Vipheaviny Chea, Jennifer G Abelin, David A Braun, Patrick A Ott, Hasmik Keshishian, Nir Hacohen, Derin B Keskin, Catherine J Wu, Steven A Carr
ABSTRAKT
MS is the most effective method to directly identify peptides presented on human leukocyte antigen (HLA) molecules. However, current standard approaches often use 500 million or more cells as input to achieve high coverage of the immunopeptidome, and therefore, these methods are not compatible with the often limited amounts of tissue available from clinical tumor samples. Here, we evaluated microscaled basic reversed-phase fractionation to separate HLA peptide samples offline followed by ion mobility coupled to LC-MS/MS for analysis. The combination of these two separation methods enabled identification of 20% to 50% more peptides compared with samples analyzed without either prior fractionation or use of ion mobility alone. We demonstrate coverage of HLA immunopeptidomes with up to 8107 distinct peptides starting with as few as 100 million cells. The increased sensitivity obtained using our methods can provide data useful to improve HLA-binding prediction algorithms as well as to enable detection of clinically relevant epitopes such as neoantigens.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis