Merck
A whole blood immunoassay using gold nanoshells.
Analytical chemistry (2003-08-16)
L R Hirsch, J B Jackson, A Lee, N J Halas, J L West
ABSTRAKT
A rapid immunoassay capable of detecting analyte within complex biological media without any sample preparation is described. This was accomplished using gold nanoshells, layered dielectric-metal nanoparticles whose optical resonance is a function of the relative size of its constituent layers. Aggregation of antibody/nanoshell conjugates with extinction spectra in the near-infrared was monitored spectroscopically in the presence of analyte. Successful detection of immunoglobulins was achieved in saline, serum, and whole blood. This system constitutes a simple immunoassay capable of detecting sub-nanogram-per-milliliter quantities of various analytes in different media within 10-30 min.