Merck
Isolation of Microvascular Endothelial Cells.
Bio-protocol (2018-06-20)
Kenneth C P Cheung, Federica M Marelli-Berg
ABSTRAKT
The vascular endothelium is essential to normal vascular homeostasis. Its dysfunction participates in various cardiovascular disorders. Murine endothelial cell culture is an important tool for cardiovascular disease research. This protocol demonstrates a quick, efficient method for the isolation of microvascular endothelial cells from murine tissues without any special equipment. To isolate endothelial cells, the lung or heart were mechanically minced and enzymatically digested with collagenase and trypsin. The single cell suspension obtained was then incubated with an anti-CD31, anti-CD105 antibody and with biotinylated isolectin B-4. The endothelial cells were harvested using magnetic bead separation with rat anti-mouse Ig- and streptavidin-conjugated microbeads. Endothelial cells were expanded and collected for subsequent analyses. The morphological and phenotypic features of these cultures remained stable over 10 passages in culture. There was no overgrowth of contaminating cells of non-endothelial origin at any stage.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Endothelial cell growth supplement from bovine pituitary, ECGS, suitable for cell culture, BioReagent