Merck
Acquired FGFR and FGF Alterations Confer Resistance to Estrogen Receptor (ER) Targeted Therapy in ER+ Metastatic Breast Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-07-30)
Pingping Mao, Ofir Cohen, Kailey J Kowalski, Justin G Kusiel, Jorge E Buendia-Buendia, Michael S Cuoco, Pedro Exman, Seth A Wander, Adrienne G Waks, Utthara Nayar, Jon Chung, Samuel Freeman, Orit Rozenblatt-Rosen, Vincent A Miller, Federica Piccioni, David E Root, Aviv Regev, Eric P Winer, Nancy U Lin, Nikhil Wagle
ABSTRAKT
To identify clinically relevant mechanisms of resistance to ER-directed therapies in ER+ breast cancer. We conducted a genome-scale functional screen spanning 10,135 genes to investigate genes whose overexpression confer resistance to selective estrogen receptor degraders. In parallel, we performed whole-exome sequencing in paired pretreatment and postresistance biopsies from 60 patients with ER+ metastatic breast cancer who had developed resistance to ER-targeted therapy. Furthermore, we performed experiments to validate resistance genes/pathways and to identify drug combinations to overcome resistance. Pathway analysis of candidate resistance genes demonstrated that the FGFR, ERBB, insulin receptor, and MAPK pathways represented key modalities of resistance. The FGFR pathway was altered via FGFR1, FGFR2, or FGF3 amplifications or FGFR2 mutations in 24 (40%) of the postresistance biopsies. In 12 of the 24 postresistance tumors exhibiting FGFR/FGF alterations, these alterations were acquired or enriched under the selective pressure of ER-directed therapy. In vitro experiments in ER+ breast cancer cells confirmed that FGFR/FGF alterations led to fulvestrant resistance as well as cross-resistance to the CDK4/6 inhibitor palbociclib. RNA sequencing of resistant cell lines demonstrated that FGFR/FGF induced resistance through ER reprogramming and activation of the MAPK pathway. The resistance phenotypes were reversed by FGFR inhibitors, a MEK inhibitor, and/or a SHP2 inhibitor. Our results suggest that FGFR pathway is a distinct mechanism of acquired resistance to ER-directed therapy that can be overcome by FGFR and/or MAPK pathway inhibitors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis