Merck
Assessment of galactose-1-phosphate uridyltransferase activity in cells and tissues.
Journal of biological methods (2021-07-15)
Megan L Brophy, John E Murphy, Robert D Bell
ABSTRAKT
Galactosemias are a family of autosomal recessive genetic disorders resulting from impaired enzymes of the Leloir pathway of galactose metabolism including galactokinase, galactose uridyltransferase, and UDP-galactose 4-epimerase that are critical for conversion of galactose into glucose-6-phosphate. To better understand pathophysiological mechanisms involved in galactosemia and develop novel therapies to address the unmet need in patients, it is important to develop reliable assays to measure the activity of the Leloir pathway enzymes. Here we describe in-depth methods for indirectly measuring galacose-1-phosphate uridyltransferase activity in cell culture and animal tissues.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Saponin, Molecular Biology, used as non-ionic surfactant
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Roche
cOmplete, Mini, Koktajl inhibitorów proteaz bez EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial