- A new kaempferol diglycoside from Datura suaveolens Humb. & Bonpl. ex. Willd.
A new kaempferol diglycoside from Datura suaveolens Humb. & Bonpl. ex. Willd.
Natural product research (2006-11-28)
A Sajeli Begum, Mahendra Sahai, Yoshinori Fujimoto, K Asai, Kathrin Schneider, Graeme Nicholson, Roderich Suessmuth
PMID17127514
ABSTRAKT
A new flavonol glycoside, kaempferol 3-O-alpha-L-arabinopyranosyl-7-O-beta-D-glucopyranoside (1), has been isolated from methanol extract of leaves of Datura suaveolens (Solanaceae), along with six other known compounds, which include kaempferol 3-O-alpha-L-arabinopyranoside (2), 3-phenyl lactic acid, 3-(3-indolyl) lactic acid, and its methyl ester, physalindicanol A and physalindicanol B. The structural elucidation of 1 and characterization of the known compounds are based on detailed spectral analysis (ESI-FTICR-MS and 2D-NMR). This is the first report of isolation of these compounds from this plant.