Merck
Impact of mucin on the anti-erosive/anti-abrasive efficacy of chitosan and/or F/Sn in enamel in vitro.
Scientific reports (2021-03-07)
Benedikt Luka, Vivien Arbter, Kathrin Sander, Andrea Duerrschnabel, Nadine Schlueter
ABSTRAKT
The application of stannous ions in combination with fluoride (F/Sn) is one of the central strategies in reducing erosive tooth wear. F/Sn efficacy can be enhanced by adding chitosan, a positively charged biopolymer. For patients with low saliva flow, this efficacy, however, is not sufficient, making further improvement desirable. This could be achieved by combining chitosan with other molecules like mucin, which together might form multilayers. This in-vitro study aimed to investigate the effect of chitosan, mucin, F/Sn and combinations thereof on enamel erosion and erosion-abrasion. Human enamel samples (n = 448, 28 groups) were cyclically eroded or eroded-abraded (10 days; 6 × 2 min erosion and 2 × 15 s/200 g abrasion per day). Samples were treated 2 × 2 min/day with solutions containing either, chitosan (50 or 500 mPas), porcine gastric mucin, F/Sn or combinations thereof after abrasive challenge. Tissue loss was measured profilometrically, interaction between hard tissue and active agents was assessed with energy dispersive spectroscopy and scanning electron microscopy. Chitosan and F/Sn showed the expected effect in reducing tissue loss under erosive and under erosive-abrasive conditions. Neither mucin alone nor the combinations with mucin showed any additional beneficial effect.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mucin from porcine stomach, Type II