Merck
Effect of glycosylation on the biochemical properties of beta-xylosidases from Aspergillus versicolor.
Journal of microbiology (Seoul, Korea) (2009-06-27)
Alexandre Favarin Somera, Marita Gimenez Pereira, Luis Henrique Souza Guimarães, Maria de Lourdes Teixeira de Moraes Polizeli, Héctor Francisco Terenzi, Rosa Prazeres Melo Furriel, João Atílio Jorge
ABSTRAKT
Aspergillus versicolor grown on xylan or xylose produces two beta-xylosidases with differences in biochemical properties and degree of glycosylation. We investigated the alterations in the biochemical properties of these beta-xylosidases after deglycosylation with Endo-H or PNGase F. After deglycosylation, both enzymes migrated faster in PAGE or SDS-PAGE exhibiting the same R(f). Temperature optimum of xylan-induced and xylose-induced beta-xylosidases was 45 degrees C and 40 degrees C, respectively, and 35 degrees C after deglycosylation. The xylan-induced enzyme was more active at acidic pH. After deglycosylation, both enzymes had the same pH optimum of 6.0. Thermal resistance at 55 degrees C showed half-life of 15 min and 9 min for xylose- and xylan-induced enzymes, respectively. After deglycosylation, both enzymes exhibited half-lives of 7.5 min. Native enzymes exhibited different responses to ions, while deglycosylated enzymes exhibited identical responses. Limited proteolysis yielded similar polypeptide profiles for the deglycosylated enzymes, suggesting a common polypeptide core with differential glycosylation apparently responsible for their biochemical and biophysical differences.