Merck
Immunohistochemical Evaluation of FGD3 Expression: A New Strong Prognostic Factor in Invasive Breast Cancer.
Cancers (2021-08-08)
Tommaso Susini, Giulia Saccardin, Irene Renda, Milo Giani, Enrico Tartarotti, Jacopo Nori, Ermanno Vanzi, Elisa Pasqualini, Simonetta Bianchi
ABSTRAKT
Among new prognostic factors for breast cancer, the most promising one seems to be FGD3 (Facio-Genital Dysplasia 3) gene, whose expression improves outcome by inhibiting cell migration. The aim of the study was to evaluate the prognostic role of FGD3 in invasive breast cancer in a series of 401 women, treated at our unit, by evaluating the expression of this gene by immunohistochemistry. Patients with high FGD3 expression showed a significantly better disease-free survival (DFS) (p < 0.001) and overall survival (OS) (p < 0.001). The prognostic value of FGD3 expression was stronger than that of classical pathologic parameters such as histological grade of differentiation, Ki-67 index and molecular subtype. By multivariate Cox analysis, FGD3 expression was confirmed as significant and independent prognostic factor, ranking second after age at diagnosis (≤40 years) for DFS (p = 0.003) and the second strongest predictor of OS, after AJCC Stage (p < 0.001). Our data suggest that inclusion of FGD3 evaluation in the routine workup of breast cancer patients may result in a more accurate stratification of the individual risk. The possibility to assess FGD3 expression by a simple and cheap technique such as immunohistochemistry may enhance the spread of its use in the clinical practice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-FGD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution