Merck
Docosahexaenoic Acid and Eicosapentaenoic Acid Inhibit the Contractile Responses of the Guinea Pig Lower Gastrointestinal Tract.
Biological & pharmaceutical bulletin (2021-08-03)
Keisuke Obara, Ayana Kawaguchi, Rikako Inaba, Mirai Kawakita, Rika Yamaguchi, Haruna Yamashita, Keyue Xu, Guanghan Ou, Fumiko Yamaki, Kento Yoshioka, Yoshio Tanaka
ABSTRAKT
Docosahexaenoic acid (DHA) and eicosapentaenoic acid (EPA) are n-3 polyunsaturated fatty acids (PUFAs), and are abundant in fish oil. These n-3 PUFAs have been reported to improve the lower gastrointestinal (LGI) disorders such as ulcerative colitis and Crohn's disease through their anti-inflammatory effects. However, there are few studies on the effect of n-3 PUFAs on motility of the LGI tract, such as the ileum and colon, the parts frequently affected by these inflammatory disorders. To elucidate the effects of DHA and EPA on the LGI tract motility, we performed comparative evaluation of their effects and linoleic acid (LA), an n-6 PUFA, on contractions in the ileal and colonic longitudinal smooth muscles (LSMs) isolated from guinea pigs. In the ileal and colonic LSMs, DHA and EPA (3 × 10-5 M each) significantly inhibited contractions induced by acetylcholine (ACh), histamine, and prostaglandin (PG) F2α (vs. control), and these effects are stronger than that of LA (3 × 10-5 M). In the colonic LSMs, DHA and EPA also significantly inhibited contractions induced by PGD2 (vs. control). In addition, DHA and EPA significantly inhibited CaCl2-induced ileal and colonic LSM contractions in Ca2+-free 80 mM-KCl solution (vs. control). Any ileal and colonic LSM contractions induced by ACh, histamine, PGF2α, and CaCl2 were completely suppressed by verapamil (10-5 M), a voltage-gated/dependent Ca2+ channel (VGCC/VDCC) inhibitor. These findings suggest that DHA and EPA could improve the abnormal contractile functions of the LGI tract associated with inflammatory diseases, partly through inhibition of VGCC/VDCC-dependent ileal and colonic LSM contractions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MK-0524, ≥98% (HPLC)