Merck
BRCA1 deficiency specific base substitution mutagenesis is dependent on translesion synthesis and regulated by 53BP1.
Nature communications (2022-01-13)
Dan Chen, Judit Z Gervai, Ádám Póti, Eszter Németh, Zoltán Szeltner, Bernadett Szikriszt, Zsolt Gyüre, Judit Zámborszky, Marta Ceccon, Fabrizio d'Adda di Fagagna, Zoltan Szallasi, Andrea L Richardson, Dávid Szüts
ABSTRAKT
Defects in BRCA1, BRCA2 and other genes of the homology-dependent DNA repair (HR) pathway cause an elevated rate of mutagenesis, eliciting specific mutation patterns including COSMIC signature SBS3. Using genome sequencing of knock-out cell lines we show that Y family translesion synthesis (TLS) polymerases contribute to the spontaneous generation of base substitution and short insertion/deletion mutations in BRCA1 deficient cells, and that TLS on DNA adducts is increased in BRCA1 and BRCA2 mutants. The inactivation of 53BP1 in BRCA1 mutant cells markedly reduces TLS-specific mutagenesis, and rescues the deficiency of template switch-mediated gene conversions in the immunoglobulin V locus of BRCA1 mutant chicken DT40 cells. 53BP1 also promotes TLS in human cellular extracts in vitro. Our results show that HR deficiency-specific mutagenesis is largely caused by TLS, and suggest a function for 53BP1 in regulating the choice between TLS and error-free template switching in replicative DNA damage bypass.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture