Merck
  • Expression of sphingosine 1-phosphate receptor 4 and sphingosine kinase 1 is associated with outcome in oestrogen receptor-negative breast cancer.
Expression of sphingosine 1-phosphate receptor 4 and sphingosine kinase 1 is associated with outcome in oestrogen receptor-negative breast cancer.
British journal of cancer (2012-03-31)
J Ohotski, J S Long, C Orange, B Elsberger, E Mallon, J Doughty, S Pyne, N J Pyne, J Edwards
ABSTRAKT
We previously reported that sphingosine 1-phosphate receptor 4 (S1P(4)) is expressed and stimulates the ERK-1/2 pathway via a human epidermal growth factor receptor 2 (HER2)-dependent mechanism in oestrogen receptor-negative (ER(-)) MDA-MB-453 breast cancer cells. Clinical relevance of S1P(4) and sphingosine kinase 1 (SK1, which catalyses the formation of S1P) was assessed in a cohort of 140 ER(-) breast tumours by immunohistochemistry (IHC) and the weighted histoscore method. Additional evidence for a functional interaction between S1P(4) and SK1 and between HER2 and SK1 was obtained using MDA-MB-453 cells. High S1P(4) expression is associated with shorter disease-free (P=0.014) and disease-specific survival (P=0.004), and was independent on multivariate analysis. In addition, patients with tumours that contain high and low levels of SK1 and S1P(4), respectively, have a significantly shorter disease-free survival (P=0.043) and disease-specific survival (P=0.033) compared with patients whose tumours contain both low S1P(4) and SK1 levels. In addition, high tumour expression of SK1 was significantly associated with shorter disease-specific survival (P=0.0001) in patients with HER2-positive tumours. Treatment of MDA-MB-453 cells with the SK1 inhibitor, SKi (2-(p-hydroxyanilino)-4-(p-chlorophenyl)thiazole) reduced the basal and S1P/S1P(4)-induced activation of ERK-1/2 and altered HER2 trafficking in these cells. These findings highlight an important role for S1P(4) and SK1 in ER(-) breast cancer progression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sphingosine kinase 2 Active human, recombinant, expressed in baculovirus, ≥80% (SDS-PAGE)