Merck
Chemoattraction of macrophages, T lymphocytes, and mast cells is evolutionarily conserved within the human alpha-defensin family.
Journal of immunology (Baltimore, Md. : 1950) (2007-09-06)
Jasmin Grigat, Afsaneh Soruri, Ulf Forssmann, Joachim Riggert, Jörg Zwirner
ABSTRAKT
Human defensins are natural peptide antibiotics. On the basis of the position and bonding of six conserved cysteine residues, they are divided into two families, designated alpha- and beta-defensins. Human alpha-defensins are expressed predominantly in neutrophils (human neutrophil peptides (HNP) 1-4) or intestinal Paneth cells (human defensins (HD) 5 and 6). Although alpha-defensins have been implicated in the pathogenesis of inflammatory bowel disease, their immunomodulatory functions are poorly understood. In the present study, HNP-1, HNP-3, and HD5 were found to be potent chemotaxins for macrophages but not dendritic cells using Galphai proteins and MAPK as signal transducers. Alpha-defensins were also chemoattractive for the human mast cell line HMC-1 but lacked, in contrast to beta-defensins, the ability to induce intracellular calcium fluxes. Furthermore, HNP-1, HNP-3, and HD5 comparably mobilized naive as well as memory T lymphocytes. Using the protein kinase C (PKC) inhibitors GF109 and Gö6976, we observed a PKC-independent functional desensitization to occur between human alpha-defensins, which suggests a common receptor for HNP-1, HNP-3, and HD5 on immune cells. This alpha-defensin receptor was subject to heterologous desensitization by the PKC activator PMA and to PKC-dependent cross-desensitization by human beta-defensins. Conversely, alpha-defensins desensitized beta-defensin-mediated migration of immune cells in a PKC-dependent manner, suggesting unique receptors for both defensin families. Taken together, our observations indicate that chemoattraction of macrophages, T lymphocytes, and mast cells represents an immunomodulatory function which is evolutionarily conserved within the human alpha-defensin family and tightly regulated by beta-defensins.