Merck
Reorganizing the protein space at the Universal Protein Resource (UniProt).
Nucleic acids research (2011-11-22)
ABSTRAKT
The mission of UniProt is to support biological research by providing a freely accessible, stable, comprehensive, fully classified, richly and accurately annotated protein sequence knowledgebase, with extensive cross-references and querying interfaces. UniProt is comprised of four major components, each optimized for different uses: the UniProt Archive, the UniProt Knowledgebase, the UniProt Reference Clusters and the UniProt Metagenomic and Environmental Sequence Database. A key development at UniProt is the provision of complete, reference and representative proteomes. UniProt is updated and distributed every 4 weeks and can be accessed online for searches or download at http://www.uniprot.org.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Uricase from Bacillus fastidiosus, lyophilized, ~9 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg