Merck
Current state of domino transplantation in Japan in terms of surgical procedures and de novo amyloid neuropathy
Amyloid : the International Journal of Experimental and Clinical Investigation : the Official Journal of the International Society of Amyloidosis (2012)
Asonuma, K., et al.