Merck
Human SND2 mediates ER targeting of GPI-anchored proteins with low hydrophobic GPI attachment signals.
FEBS letters (2021-04-11)
Jing Yang, Tetsuya Hirata, Yi-Shi Liu, Xin-Yu Guo, Xiao-Dong Gao, Taroh Kinoshita, Morihisa Fujita
ABSTRAKT
Over 100 glycosylphosphatidylinositol-anchored proteins (GPI-APs) are encoded in the mammalian genome. It is not well understood how these proteins are targeted and translocated to the endoplasmic reticulum (ER). Here, we reveal that many GPI-APs, such as CD59, CD55, and CD109, utilize human SND2 (hSND2)-dependent ER targeting machinery. We also found that signal recognition particle receptors seem to cooperate with hSND2 to target GPI-APs to the ER. Both the N-terminal signal sequence and C-terminal GPI attachment signal of GPI-APs contribute to ER targeting via the hSND2-dependent pathway. Particularly, the hydrophobicity of the C-terminal GPI attachment signal acts as the determinant of hSND2 dependency. Our results explain the route and mechanism of the ER targeting of GPI-APs in mammalian cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
d-Desthiobiotin, ≥98% (TLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse