Merck
Orphan nuclear receptor NR4A1 suppresses hyperhomocysteinemia-induced hepatic steatosis in vitro and in vivo.
FEBS letters (2019-04-12)
Hongjin Liang, Xina Xie, Xuhong Song, Meihui Huang, Ting Su, Xiaolan Chang, Bin Liang, Dongyang Huang
ABSTRAKT
Homocysteine (Hcy) is associated with nonalcoholic fatty liver disease (NAFLD). orphan nuclear receptor subfamily 4 group A member 1 (NR4A1) is involved in hepatic lipid metabolism. However, the potential role of NR4A1 in Hcy-associated NAFLD remains elusive. We aimed to elucidate the regulation of NR4A1 and its significance in Hcy-induced NAFLD. Hcy induced steatosis and elevated the expression of CD36 and FATP2 in HepG2 cells. Furthermore, Hcy enhanced p300 and decreased HDAC7 recruitment to the NR4A1 promoter, resulting in histone H3K27 hyperacetylation and NR4A1 upregulation. Moreover, NR4A1 depletion not only mimicked but also exaggerated the effects of Hcy on steatosis, whereas NR4A1 agonist Cytosporone B (CsnB) blocked Hcy-induced steatosis. In hyperhomocysteinemia (HHcy) mice, CsnB attenuated HHcy-induced hepatic steatosis. Thus, Hcy transiently and rapidly induces NR4A1 expression to reduce Hcy-induced steatosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys4) - przeciwciało z walidacją ChIP i zestaw primerów, królicze monoklonalne, from rabbit
Sigma-Aldrich
ChIPAb+ Dimetylo-Histon H3 (Lys9) - przeciwciało i zestaw primerów zweryfikowane pod kątem ChIP, serum, from rabbit
Sigma-Aldrich
ChIPAb+ Acetylo-Histone H3 (Lys27) - przeciwciało i zestaw primerów zweryfikowane pod kątem ChIP, clone CMA309, from mouse
Sigma-Aldrich
ChIPAb+ HDAC1 - przeciwciało z walidacją ChIP i zestaw starterów, culture supernatant, from mouse