Merck
  • Supplementing cultured human myotubes with hibernating bear serum results in increased protein content by modulating Akt/FOXO3a signaling.
Supplementing cultured human myotubes with hibernating bear serum results in increased protein content by modulating Akt/FOXO3a signaling.
PloS one (2022-01-26)
Mitsunori Miyazaki, Michito Shimozuru, Toshio Tsubota
ABSTRAKT
Hibernating bears remain in their dens for 5-7 months during winter and survive without eating or drinking while staying inactive. However, they maintain their physical functions with minimal skeletal muscle atrophy and metabolic dysfunction. In bears, resistance to skeletal muscle atrophy during hibernation is likely mediated by seasonally altered systemic factors that are independent of neuromuscular activity. To determine whether there are components in bear serum that regulate protein and energy metabolism, differentiated human skeletal muscle cells were treated with bear serum (5% in DMEM/Ham's F-12, 24 h) collected during active summer (July) and hibernating winter (February) periods. The serum samples were collected from the same individual bears (Ursus thibetanus japonicus, n = 7 in each season). Total protein content in cultured skeletal muscle cells was significantly increased following a 24 h treatment with hibernating bear serum. Although the protein synthesis rate was not altered, the expression of MuRF1 protein, a muscle-specific E3 ubiquitin ligase was significantly decreased along with a concomitant activation of Akt/FOXO3a signaling. Increased levels of insulin-like growth factor-1 (IGF-1) were also observed in hibernating bear serum. These observations suggest that protein metabolism in cultured human myotubes may be altered when incubated with hibernating bear serum, with a significant increase in serum IGF-1 and diminished MuRF1 expression, a potential target of Akt/FOXO3a signaling. A protein sparing phenotype in cultured muscle cells by treatment with hibernating bear serum holds potential for the development of methods to prevent human muscle atrophy and related disorders.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse