Merck
scGR-seq: Integrated analysis of glycan and RNA in single cells.
STAR protocols (2022-03-05)
Haruki Odaka, Haruka Ozaki, Hiroaki Tateno
ABSTRAKT
Glycans are structurally diverse molecules found on the surface of living cells. The protocol details a system developed for combined analysis of glycan and RNA in single cells (scGR-seq) using human induced pluripotent stem cells (hiPSCs) and hiPSC-derived neural progenitor cells (NPCs). scGR-seq consists of DNA-barcoded lectin-based glycan profiling by sequencing (scGlycan-seq) and single-cell transcriptome profiling (scRNA-seq). scGR-seq will be an essential technique to delineate the cellular heterogeneity of glycans across multicellular systems. For complete details on the use and execution of this profile, please refer to Minoshima et al. (2021).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse