Merck
BECN1 modulates hematopoietic stem cells by targeting Caspase-3-GSDME-mediated pyroptosis.
Blood science (Baltimore, Md.) (2020-07-25)
Xiuxiu Yang, Liang Ge, Jianwei Wang
ABSTRAKT
Hematopoietic stem cells (HSCs) maintain the blood system throughout the lifespan. However, the molecular mechanism maintaining HSC character remains not fully understood. In this study, we observed that the targeted deletion of Becn1 disrupts the blood system and impairs the reconstitution capacity of HSCs. Interestingly, Becn1 deletion did not lead to dysfunction of autophagy in HSCs, indicating a non-classical role of BECN1 in regulating HSCs function. While we observed the increase of Caspase-3-GSDME-mediated pyroptosis in Becn1 deficient hematopoietic stem and progenitor cells. Forced expression of the full-length GSDME compromises the function of HSCs. In brief, we identified a novel role of Becn1 in modulating HSCs by regulating pyroptosis, but not through autophagy. This study provides a new link between BECN1-Caspase-3-GSDME signaling and HSC maintenance.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence