Merck
PAX2 induces vascular-like structures in normal ovarian cells and ovarian cancer.
Experimental and therapeutic medicine (2022-05-24)
Kholoud Alwosaibai, Ensaf M Al-Hujaily, Salmah Alamri, Salim Ghandorah, Kenneth Garson, Barbara C Vanderhyden
ABSTRAKT
In adult tissue, the paired box 2 (PAX2) protein is expressed in healthy oviductal, but not normal ovarian surface epithelial cells. PAX2 is expressed in a subset of cases of serous ovarian carcinoma; however, the role of PAX2 in the initiation and progression of ovarian cancer remains unknown. The aim of the present study was to determine the biological effects of PAX2 expression in normal and cancerous epithelial cells. By culturing the normal and cancerous ovarian cells that express PAX2 in 3D culture and staining the cells with vasculogenic mimicry markers such as CD31 and PAS, it was shown that PAX2 overexpression in both normal and cancerous ovarian epithelial cells induced formation of vascular-like structures both in vitro and in vivo. These results indicated a potential role of PAX2 in ovarian cancer progression by increasing the presence of vascular-like structures to promote the supply of nutrients to tumor cells and facilitate cancer cell proliferation and invasion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Kozie przeciwciało anty-Mysie IgG, koniugat HRP, Upstate®, from goat