Merck
  • Predictors of efficacy of corticosteroid switching from abiraterone plus prednisone to dexamethasone in patients with metastatic castration-resistant prostate cancer.
Predictors of efficacy of corticosteroid switching from abiraterone plus prednisone to dexamethasone in patients with metastatic castration-resistant prostate cancer.
Asian journal of andrology (2021-08-13)
Yu-Chao Ni, Jin-Ge Zhao, Meng-Ni Zhang, Yi-Jun Zhang, Zhen-Yu Yang, Ni Chen, Jun-Ru Chen, Peng-Fei Shen, Guang-Xi Sun, Xing-Ming Zhang, Yong-Hong Li, Hao Zeng
ABSTRAKT
Corticosteroid switching can reverse abiraterone resistance in some patients with metastatic castration-resistant prostate cancer (mCRPC). Here, we investigated the potential biomarkers for predicting the efficacy of corticosteroid switching during treatment with abiraterone acetate (AA). We retrospectively analyzed 101 mCRPC patients receiving corticosteroid switching from West China Hospital and Sun Yat-Sen University Cancer Center between January 2016 and December 2018. All cases received AA plus prednisone as first-line therapy during mCRPC. Primary end points were biochemical progression-free survival (bPFS) and overall survival (OS). The risk groups were defined based on multivariate analysis. A total of 42 (41.6%) and 25 (24.8%) patients achieved 30% and 50% decline in prostate-specific antigen (PSA), respectively, after corticosteroid switching. The median bPFS and median OS on AA plus dexamethasone were 4.9 (95% confidence interval [CI]: 3.7-6.0) months and 18.8 (95% CI: 16.2-30.2) months, respectively. Aldo-keto reductase family 1 member C3 (AKR1C3) expression (hazard ratio [HR]: 2.15, 95% Cl: 1.22-3.80, P = 0.008) and baseline serum alkaline phosphatase (ALP; HR: 4.95, 95% Cl: 2.40-10.19, P < 0.001) were independent predictors of efficacy before corticosteroid switching in the multivariate analysis of bPFS. Only baseline serum ALP >160 IU l-1 (HR: 3.41, 95% Cl: 1.57-7.38, P = 0.002) together with PSA level at switch ≥50 ng ml-1 (HR: 2.59, 95% Cl: 1.22-5.47, P = 0.013) independently predicted poorer OS. Based on the predictive factors in multivariate analysis, we developed two risk stratification tools to select candidates for corticosteroid switching. Detection of serum ALP level, PSA level, and tissue AKR1C3 expression in mCRPC patients could help make clinical decisions for corticosteroid switching.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-AKR1C3 antibody, Mouse monoclonal, clone NP6.G6.A6, purified from hybridoma cell culture