Merck
Laminar Flow on Endothelial Cells Suppresses eNOS O-GlcNAcylation to Promote eNOS Activity.
Circulation research (2021-10-05)
Sarah E Basehore, Samantha Bohlman, Callie Weber, Swathi Swaminathan, Yuji Zhang, Cholsoon Jang, Zoltan Arany, Alisa Morss Clyne
ABSTRAKT
[Figure: see text].
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline