Merck
  • Multicomponent Synthesis, Binding Mode, and Structure-Activity Relationship of Selective Histone Deacetylase 6 (HDAC6) Inhibitors with Bifurcated Capping Groups.
Multicomponent Synthesis, Binding Mode, and Structure-Activity Relationship of Selective Histone Deacetylase 6 (HDAC6) Inhibitors with Bifurcated Capping Groups.
Journal of medicinal chemistry (2020-08-18)
Nina Reßing, Melf Sönnichsen, Jeremy D Osko, Andrea Schöler, Julian Schliehe-Diecks, Alexander Skerhut, Arndt Borkhardt, Julia Hauer, Matthias U Kassack, David W Christianson, Sanil Bhatia, Finn K Hansen
ABSTRAKT
Histone deacetylase 6 (HDAC6) is an emerging target for the treatment of cancer, neurodegenerative diseases, inflammation, and other diseases. Here, we present the multicomponent synthesis and structure-activity relationship of a series of tetrazole-based HDAC6 inhibitors. We discovered the hit compound NR-160 by investigating the inhibition of recombinant HDAC enzymes and protein acetylation. A cocrystal structure of HDAC6 complexed with NR-160 disclosed that the steric complementarity of the bifurcated capping group of NR-160 to the L1 and L2 loop pockets may be responsible for its HDAC6-selective inhibition. While NR-160 displayed only low cytotoxicity as a single agent against leukemia cell lines, it augmented the apoptosis induction of the proteasome inhibitor bortezomib in combination experiments significantly. Furthermore, a combinatorial high-throughput drug screen revealed significantly enhanced cytotoxicity when NR-160 was used in combination with epirubicin and daunorubicin. The synergistic effect in combination with bortezomib and anthracyclines highlights the potential of NR-160 in combination therapies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
NR-160, ≥98% (HPLC)