Merck
  • Lysosomal rupture induced by structurally distinct chitosans either promotes a type 1 IFN response or activates the inflammasome in macrophages.
Lysosomal rupture induced by structurally distinct chitosans either promotes a type 1 IFN response or activates the inflammasome in macrophages.
Biomaterials (2017-03-25)
David Fong, Pascal Grégoire-Gélinas, Alexandre P Cheng, Tal Mezheritsky, Marc Lavertu, Sachiko Sato, Caroline D Hoemann
ABSTRAKT
Chitosan is a family of glucosamine and N-acetyl glucosamine polysaccharides with poorly understood immune modulating properties. Here, functional U937 macrophage responses were analyzed in response to a novel library of twenty chitosans with controlled degree of deacetylation (DDA, 60-98%), molecular weight (1 to >100 kDa), and acetylation pattern (block vs. random). Specific chitosan preparations (10 or 190 kDa 80% block DDA and 3, 5, or 10 kDa 98% DDA) either induced macrophages to release CXCL10 and IL-1ra at 5-50 μg/mL, or activated the inflammasome to release IL-1β and PGE2 at 50-150 μg/mL. Chitosan induction of these factors required lysosomal acidification. CXCL10 production was preceded by lysosomal rupture as shown by time-dependent co-localization of galectin-3 and chitosan and slowed autophagy flux, and specifically depended on IFN-β paracrine activity and STAT-2 activation that could be suppressed by PGE2. Chitosan induced a type I IFN paracrine response or inflammasome response depending on the extent of lysosomal rupture and cytosolic foreign body invasion. This study identifies the structural motifs that lead to chitosan-driven cytokine responses in macrophages and indicates that lysosomal rupture is a key mechanism that determines the endogenous release of either IL-1ra or IL-1β.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-STAT2, Upstate®, from rabbit