Merck
  • Exosome miR-23a-3p from Osteoblast Alleviates Spinal Cord Ischemia/Reperfusion Injury by Down-Regulating KLF3-Activated CCNL2 Transcription.
Exosome miR-23a-3p from Osteoblast Alleviates Spinal Cord Ischemia/Reperfusion Injury by Down-Regulating KLF3-Activated CCNL2 Transcription.
Developmental neuroscience (2021-12-23)
Cheng Wu, Qinghua Zhu, Yi Yao, Zhaoyang Shi, Chaojie Jin, Leilei Chen
ABSTRAKT
Spinal cord ischemia/reperfusion injury (SCIRI) is usually caused by spinal surgery or aortic aneurysm surgery and can eventually lead to paralysis or paraplegia and neurological dysfunction. Exosomes are considered as one of the most promising therapeutic strategies for SCIRI as they can pass the blood-spinal barrier. Previous studies have proved that exosomes secreted by osteocytes have a certain slowing effect on SCIRI. We aimed to explore the effect of osteoblast secreted exosomes on SCIRI. First, neurons and osteoblasts were co-cultured under different conditions. GEO database was utilized to detect the expression of miR-23a-3p in osteoblast exosomes. SCIRI cells were treated with exosomes, and the detection was taken to prove whether miR-23a-3p could slow the progression of SCIRI. Downstream gene and the potential regulatory mechanism were explored through database and functional experiments. MiR-23a-3p was highly expressed in exosomes and it slowed down the process of SCIRI. Downstream mRNA KLF3 could bind to miR-23a-3p and was highly expressed in IRI. Moreover, CCNL2 was regulated by KLF3 and was highly expressed in IRI. Rescue experiments verified that miR-23a-3p suppressed the transcription of CCNL2 by targeting KLF3. Exosome miR-23a-3p from osteoblast alleviates SCIRI by down-regulating KLF3-activated CCNL2 transcription.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
EZ-Magna ChIP® A - Zestaw do immunoprecypitacji chromatyny, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.