Merck
Fetal bovine serum-a cell culture dilemma.
Science (New York, N.Y.) (2022-01-14)
Jan van der Valk
ABSTRAKT
Ethical and possible reproducibility issues arise when using fetal bovine serum in cell culture media.