Merck
Imprinted SARS-CoV-2-specific memory lymphocytes define hybrid immunity.
Cell (2022-04-13)
Lauren B Rodda, Peter A Morawski, Kurt B Pruner, Mitchell L Fahning, Christian A Howard, Nicholas Franko, Jennifer Logue, Julie Eggenberger, Caleb Stokes, Inah Golez, Malika Hale, Michael Gale, Helen Y Chu, Daniel J Campbell, Marion Pepper
ABSTRAKT
Immune memory is tailored by cues that lymphocytes perceive during priming. The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) pandemic created a situation in which nascent memory could be tracked through additional antigen exposures. Both SARS-CoV-2 infection and vaccination induce multifaceted, functional immune memory, but together, they engender improved protection from disease, termed hybrid immunity. We therefore investigated how vaccine-induced memory is shaped by previous infection. We found that following vaccination, previously infected individuals generated more SARS-CoV-2 RBD-specific memory B cells and variant-neutralizing antibodies and a distinct population of IFN-γ and IL-10-expressing memory SARS-CoV-2 spike-specific CD4+ T cells than previously naive individuals. Although additional vaccination could increase humoral memory in previously naive individuals, it did not recapitulate the distinct CD4+ T cell cytokine profile observed in previously infected subjects. Thus, imprinted features of SARS-CoV-2-specific memory lymphocytes define hybrid immunity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder