Merck
Rac1 promotes kidney collecting duct integrity by limiting actomyosin activity.
The Journal of cell biology (2021-10-15)
Fabian Bock, Bertha C Elias, Xinyu Dong, Diptiben V Parekh, Glenda Mernaugh, Olga M Viquez, Anjana Hassan, Venkateswara Rao Amara, Jiageng Liu, Kyle L Brown, Andrew S Terker, Manuel Chiusa, Leslie S Gewin, Agnes B Fogo, Cord H Brakebusch, Ambra Pozzi, Roy Zent
ABSTRAKT
A polarized collecting duct (CD), formed from the branching ureteric bud (UB), is a prerequisite for an intact kidney. The small Rho GTPase Rac1 is critical for actin cytoskeletal regulation. We investigated the role of Rac1 in the kidney collecting system by selectively deleting it in mice at the initiation of UB development. The mice exhibited only a mild developmental phenotype; however, with aging, the CD developed a disruption of epithelial integrity and function. Despite intact integrin signaling, Rac1-null CD cells had profound adhesion and polarity abnormalities that were independent of the major downstream Rac1 effector, Pak1. These cells did however have a defect in the WAVE2-Arp2/3 actin nucleation and polymerization apparatus, resulting in actomyosin hyperactivity. The epithelial defects were reversible with direct myosin II inhibition. Furthermore, Rac1 controlled lateral membrane height and overall epithelial morphology by maintaining lateral F-actin and restricting actomyosin. Thus, Rac1 promotes CD epithelial integrity and morphology by restricting actomyosin via Arp2/3-dependent cytoskeletal branching.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholera Toxin B subunit, FITC conjugate, lyophilized powder
Sigma-Aldrich
Zestaw do testu aktywacji Rho, PP1/PP2A Toolbox for the selective in vitro dephosphorylation of proteins.
Sigma-Aldrich
Zestaw do testu aktywacji Rac1/Cdc42, The Rac1/Cdc42 Activation Assay provides an effective method for detecting Rac & Cdc42 activity in cell lysates.