Merck
Effect of selective IK,ACh inhibition by XAF-1407 in an equine model of tachypacing-induced persistent atrial fibrillation.
British journal of pharmacology (2020-05-22)
Merle Friederike Fenner, Helena Carstensen, Sarah Dalgas Nissen, Eva Melis Hesselkilde, Christine Scott Lunddahl, Maja Adler Hess Jensen, Ameli Victoria Loft-Andersen, Stefan Michael Sattler, Pyotr Platonov, Said El-Haou, Claire Jackson, Raymond Tang, Robert Kirby, John Ford, Ulrich Schotten, James Milnes, Ulrik Svane Sørensen, Thomas Jespersen, Rikke Buhl
ABSTRAKT
Inhibition of the G-protein gated ACh-activated inward rectifier potassium current, IK,ACh may be an effective atrial selective treatment strategy for atrial fibrillation (AF). Therefore, the anti-arrhythmic and electrophysiological properties of a novel putatively potent and highly specific IK,ACh inhibitor, XAF-1407 (3-methyl-1-[5-phenyl-4-[4-(2-pyrrolidin-1-ylethoxymethyl)-1-piperidyl]thieno[2,3-d]pyrimidin-6-yl]azetidin-3-ol), were characterised for the first time in vitro and investigated in horses with persistent AF. The pharmacological ion channel profile of XAF-1407 was investigated using cell lines expressing relevant ion channels. In addition, eleven horses were implanted with implantable cardioverter defibrillators enabling atrial tachypacing into self-sustained AF. The electrophysiological effects of XAF-1407 were investigated after serial cardioversions over a period of 1 month. Cardioversion success, drug-induced changes of atrial tissue refractoriness, and ventricular electrophysiology were assessed at baseline (day 0) and days 3, 5, 11, 17, and 29 after AF induction. XAF-1407 potently and selectively inhibited Kir 3.1/3.4 and Kir 3.4/3.4, underlying the IK,ACh current. XAF-1407 treatment in horses prolonged atrial effective refractory period as well as decreased atrial fibrillatory rate significantly (~20%) and successfully cardioverted AF, although with a decreasing efficacy over time. XAF-1407 shortened atrioventricular-nodal refractoriness, without effect on QRS duration. QTc prolongation (4%) within 15 min of drug infusion was observed, however, without any evidence of ventricular arrhythmia. XAF-1407 efficiently cardioverted sustained tachypacing-induced AF of short duration in horses without notable side effects. This supports IK,ACh inhibition as a potentially safe treatment of paroxysmal AF in horses, suggesting potential clinical value for other species including humans.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
XAF-1407, ≥98% (HPLC)