Merck
Endogenous spacing enables co-processing of microRNAs and efficient combinatorial RNAi.
Cell reports methods (2022-07-27)
Alexandra M Amen, Ryan M Loughran, Chun-Hao Huang, Rachel J Lew, Archna Ravi, Yuanzhe Guan, Emma M Schatoff, Lukas E Dow, Brooke M Emerling, Christof Fellmann
ABSTRAKT
We present Multi-miR, a microRNA-embedded shRNA system modeled after endogenous microRNA clusters that enables simultaneous expression of up to three or four short hairpin RNAs (shRNAs) from a single promoter without loss of activity, enabling robust combinatorial RNA interference (RNAi). We further developed complementary all-in-one vectors that are over one log-scale more sensitive to doxycycline-mediated activation in vitro than previous methods and resistant to shRNA inactivation in vivo. We demonstrate the utility of this system for intracranial expression of shRNAs in a glioblastoma model. Additionally, we leverage this platform to target the redundant RAF signaling node in a mouse model of KRAS-mutant cancer and show that robust combinatorial synthetic lethality efficiently abolishes tumor growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture